Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALX4 Antibody (OTI6B3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272123F
Description
ALX4 Monoclonal specifically detects ALX4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ALX4 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
ALX4 | |
Full length recombinant protein of human ALX4 (NP_068745) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI6B3 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ALX homeobox 4, FND2, homeobox protein aristaless-like 4, homeodomain transcription factor ALX4, KIAA1788 | |
Mouse | |
Affinity Purified | |
Transcription Factors and Regulators | |
60529.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction