Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALY Monoclonal Antibody (11G5), FITC
GREENER_CHOICE

Catalog No. PIMA546554
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546554 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA546554 Supplier Invitrogen™ Supplier No. MA546554
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Western blot detects a band of approximately 32 kDa (predicted molecular weight: 27 kDa).

The protein encoded by this gene is a heat stable, nuclear protein and functions as a molecular chaperone. It is thought to regulate dimerization, DNA binding, and transcriptional activity of basic region-leucine zipper (bZIP) proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALY
Applications ChIP Assay, ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 11G5
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 0.09% sodium azide
Gene ALYREF
Gene Accession No. Q86V81
Gene Alias ally of AML-1 and LEF-1; ALY; ALY/REF; Aly/REF export factor; Aly/REF export factor L homeolog; aly/REF export factor-A; alyref; alyref.L; alyref-a; BEF; bZIP enhancing factor; bZIP-enhancing factor BEF; hypothetical protein LOC537706; REF; Ref1; REF1-I; Refbp1; RNA and export factor binding protein 1; RNA and export factor-binding protein 1; Tcra enhancer-binding factor interacting protein 1; THO complex 4; THO complex subunit 4; THO complex subunit 4-A; tho4; Tho4-A; THOC4; thoc4-a; transcriptional coactivator Aly/REF; XELAEV_18043750mg; zgc:171753
Gene Symbols ALYREF
Host Species Mouse
Immunogen Full length recombinant human ALY/Ref fusion protein.
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10189
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.