Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542286
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA542286 100 μL
1 options

Catalog No. PIPA542286

Supplier: Invitrogen™ PA542286

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GGGPIRNRPA IARGAAGGGG RNRPAPYSRP KQLPDKWQHD LFDSGFGGGA Sequence homology: Cow: 100%; Dog: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.

The protein encoded by this gene is a heat stable, nuclear protein and functions as a molecular chaperone. It is thought to regulate dimerization, DNA binding, and transcriptional activity of basic region-leucine zipper (bZIP) proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALY
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ALYREF
Gene Accession No. Q86V81
Gene Alias ally of AML-1 and LEF-1; ALY; ALY/REF; Aly/REF export factor; Aly/REF export factor L homeolog; aly/REF export factor-A; alyref; alyref.L; alyref-a; BEF; bZIP enhancing factor; bZIP-enhancing factor BEF; hypothetical protein LOC537706; REF; Ref1; REF1-I; Refbp1; RNA and export factor binding protein 1; RNA and export factor-binding protein 1; Tcra enhancer-binding factor interacting protein 1; THO complex 4; THO complex subunit 4; THO complex subunit 4-A; tho4; Tho4-A; THOC4; thoc4-a; transcriptional coactivator Aly/REF; XELAEV_18043750mg; zgc:171753
Gene Symbols ALYREF
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human THOC4 (aa 51-100).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10189
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.