Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AM 0902
GREENER_CHOICESDP

Catalog No. 591450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-145-0 50 mg
59-141-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-145-0 Supplier Tocris Bioscience™ Supplier No. 5914/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective TRPA1 antagonist

AM 0902 is a potent and selective TRPA1 antagonist (IC50 = 0.02 μM for human TRPA1). Exhibits no siginificant activity against human TRPV1 or TRPV4, and rat TRPV1, TRPV3 or TRPM8. Exhibits efficacy in a rat pain model. Orally bioavailable.

Specifications

Chemical Name or Material 1-[[3-[2-(4-Chlorophenyl)ethyl]-1,2,4-oxadiazol-5-yl]methyl]-1,7-dihydro-7-methyl-6H-purin-6-one
CAS 1883711-97-4
Assay Percent Range >98%
Quantity 50 mg
Molecular Formula C17H15ClN6O2
Formula Weight Observed MW: 370.79
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.