Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AM 251
SDP

Catalog No. 111750 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-175-0 50 mg
11-171 1 mg
11-171-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 11-175-0 Supplier R&D Systems™ Supplier No. 1117/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent CB1 antagonist; also GPR55 agonist

Potent CB1 receptor antagonist (IC50 = 8 nM, Ki = 7.49 nM) that displays 306-fold selectivity over CB2 receptors. Also potent GPR55 agonist (EC50 = 39 nM) and μ-opioid receptor antagonist (Ki = 251 nM).

Fluorescent Form also available.

Chemical Identifiers

CAS 183232-66-8
Molecular Formula C22H21Cl2IN4O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 555.24
Purity >99%
Molecular Weight (g/mol) 555.241
Chemical Name or Material AM 251
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.