Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AM 4113
GREENER_CHOICESDP

Catalog No. 619350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-935-0 50 mg
61-931-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-935-0 Supplier Tocris Bioscience™ Supplier No. 6193/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective, high affinity CB1 antagonist

AM 4113 is a high affinity and selective CB1 antagonist (Kd = 0.89 nM). Exhibits 100-fold selectivity for CB1 over CB2. Attenuates CB1 agonist AM 411-induced locomotor suppression and reduces food intake in vivo, but does not induce signs of nausea.

Specifications

Chemical Name or Material 5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide
CAS 614726-85-1
Assay Percent Range >98%
Quantity 50 mg
Molecular Formula C17H12Cl3N3O
Formula Weight Observed MW: 380.66
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.