Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AM 580
GREENER_CHOICESDP

Catalog No. 076050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-605-0 50 mg
07-601-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 07-605-0 Supplier R&D Systems™ Supplier No. 0760/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Retinoic acid analog; RARα agonist

AM 580 is an analog of retinoic acid that acts as a selective RARα agonist (EC50 values are 0.3, 8.6 and 13 nM for RARα, RARβ and RARγ respectively). Significantly induces IL-4, IL-5 and IL-13 and inhibits IL-12 and IFNγ synthesis, and induces cell differentiation with over 7 times the activity of retinoic acid in vitro.

AM 580 synthesized to Ancillary Material Grade also available.

Chemical Identifiers

CAS 102121-60-8
Molecular Formula C22H25NO3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 351.44
Purity >98%
Molecular Weight (g/mol) 351.45
Chemical Name or Material AM 580
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.