Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AM 80
GREENER_CHOICESDP

Catalog No. 350750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-075-0 50 mg
35-071-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-075-0 Supplier R&D Systems™ Supplier No. 3507/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. RARα agonist; anticancer agent

Retinoic acid receptor α (RARα) agonist that induces differentiation (ED50 = 0.79 nM) and apoptosis of HL-60 cells in vitro. Exhibits antiproliferative effects against a variety of human tumor cells lines (mean values of 35, 40 and 60% growth inhibition at 0.1, 1 and 10 μM respectively) and displays anticancer activity against acute promyelocytic leukemia in vivo.

Chemical Identifiers

CAS 94497-51-5
Molecular Formula C22H25NO3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 351.44
Purity >99%
Molecular Weight (g/mol) 351.45
Chemical Name or Material AM 80
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.