Learn More
Invitrogen™ AMAC1L2 Polyclonal Antibody

Description
AMAC1L2 Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | AMAC1L2 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | SLC35G5 |
| Gene Accession No. | Q96KT7 |
| Gene Alias | acyl-malonyl condensing enzyme 1-like 2; acyl-malonyl-condensing enzyme 1-like protein 2; AMAC; AMAC1L2; protein AMAC1L2; SLC35G5; Solute carrier family 35 member G5; solute carrier family 35, member G5 |
| Gene Symbols | SLC35G5 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.