Learn More
Invitrogen™ AMACR Polyclonal Antibody

Description
The PA1-31262 immunogen is: Synthetic peptide: TDACVTPVLT FEEWHHD(C), corresponding to C Terminal amino acids 290-307 of Human AMCR, the peptide was modified post-synthesis.
Specifications
Specifications
| Antigen | AMACR |
| Applications | ELISA, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.75 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Tris glycine with 0.5% BSA, 30% glycerol and 0.02% Sodium azide; pH 7.4-7.9 |
| Gene | AMACR |
| Gene Accession No. | O09174, Q9UHK6 |
| Gene Alias | 2-arylpropionyl-CoA epimerase; 2-methylacyl-CoA racemase; Alpha-methylacyl-CoA racemase; alpha-methylacyl-Coenzyme A racemase; Amacr; AMACRD; amcr; CBAS4; Da1-8; Macr1; Marc1; OTTHUMP00000219978; OTTHUMP00000219979; RACE; RM |
| Gene Symbols | AMACR |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.