Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMACR Rabbit anti-Human, Unconjugated, Clone: RM349, Abnova™

Catalog No. 89339197 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-197 Supplier Abnova Corporation Supplier No. MAB21980
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

AMACR Monoclonal antibody specifically detects AMACR in Human samples. It is validated for WB-Ce, IHC-P.

Specifications

Antigen AMACR
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM349
Conjugate Unconjugated
Description alpha-methylacyl-CoA racemase
Gene AMACR
Gene Alias RACE
Host Species Rabbit
Immunogen A peptide corresponding to N-terminus of human AMACR
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 23600
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.