Learn More
AMACR Recombinant Rabbit Monoclonal Antibody (13H4), Zeta
Rabbit Recombinant Monoclonal Antibody Cocktail
$878.59
Specifications
| Antigen | AMACR |
|---|---|
| Clone | 13H4 |
| Concentration | 100 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-221-5762
|
ZETA CORPORATION
Z2001RL |
1 mL |
Each of 1 for $878.59
|
|
|||||
Description
A recommended positive control tissue for this product is Prostate CA, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.
AMACR Recombinant Monoclonal antibody specifically detects AMACR in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| AMACR | |
| 100 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| tris with BSA, NP-40 and <0.1% sodium azide | |
| 2-arylpropionyl-CoA epimerase; 2-methylacyl-CoA racemase; Alpha-methylacyl-CoA racemase; alpha-methylacyl-Coenzyme A racemase; Amacr; AMACRD; amcr; CBAS4; Da1-8; Macr1; Marc1; OTTHUMP00000219978; OTTHUMP00000219979; RACE; RM | |
| Human AMACR (P504S) polypeptide | |
| Primary | |
| 2-8°C | |
| AMACR |
| 13H4 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9UHK6 | |
| 23600 | |
| IgG | |
| Protein A | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.