Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Amaranth Monoclonal Antibody (3E4)
GREENER_CHOICE

Catalog No. PIMA549738
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549738 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549738 Supplier Invitrogen™ Supplier No. MA549738
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Amaranthin is a gene target that is involved in the regulation of cell growth and differentiation. This gene is found in many different types of organisms, including plants, animals, and humans. Amaranthin is particularly important in the development of the nervous system, where it plays a crucial role in the formation of neural circuits. Mutations in the Amaranthin gene have been linked to a number of different diseases and disorders, including cancer and neurodegenerative diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amaranth
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3E4
Concentration 2.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of AMAT hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.