Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Amber Reagent Bottle , Narrow Mouth with Acid Proof Polypropylene Stopper
SCI_ED

Catalog No. S27974 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
2000 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S27974 60 mL
S27975 125 mL
S27976 250 mL
S27977 500 mL
S27978 1000 mL
S27979 2000 mL
S27973 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. S27974 Supplier Eisco™ Supplier No. CH0161B
Add to Cart
Add to Cart

Narrow Mouth with Acid Proof Polypropylene Stopper. Made of high quality borosilicate glass.

Specifications

Capacity 30 mL
Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Amber
Description Socket Size: 14/23
For Use With (Application) To contain chemicals in liquid or powder form
Material Borosilicate Glass
Length (Metric) 152.4 mm
Quantity 1
Sterility Non-sterile
Standard Taper Joint 14/23
Weight (Metric) 226.79 g
Width (Metric) 101.6 mm
Closure Color White
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow
Shape Cylindrical
Height (Metric) 10.16 cm
Product Type Reagent Bottle
For Use With (Equipment) Acid
Height (English) 4 in.
Chemical Label Unlabeled
Closure Material Polypropylene
Closure Size 14/23
Closure Type Stopper
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.