Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Amber Glass Graduated Reagent Bottles with Screw Caps and Drip-Free Pouring Rings

Shop All Supertek Scientific Products


Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Description
- In conformity to DIN ISO 4796-1 Standard
- Printed in white color
- These bottles are mechanically strong and chemical resistant
- Provided with a pouring ring for drip-free operation
- The screw caps and pouring ring are made of PP
- The bottles screw caps and pouring rings can be sterilized and are autoclavable
- 100 mL to 1000 mL bottles have uniform GL 45 thread
Specifications
Specifications
| Capacity | 500 mL |
| Capacity (Metric) | 500 mL |
| Capacity (English) | 16.9 oz. |
| Color | Amber |
| For Use With (Application) | Storage |
| Includes | Drip-Free Pouring Rings |
| Material | Borosilicate Glass (3.3) |
| Quantity | 10 Pk. |
| Sterility | Sterile |
| Certifications/Compliance | DIN ISO 4796-1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction