Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AmberChrom™, 50WX2, 50-100 mesh, H-form, Thermo Scientific™
Click to view available options
Quantity:
100 g
500 g
Specifications
Specifications
| Physical Form | Beads |
| CAS Min % | 18%, 74% |
| CAS Max % | 26%, 82% |
| Ion Exchange Capacity | ≥0.6 eq/L (H form) |
| Particle Size | 50 to 100 mesh, ≤15 % (< 210 μm), ≤15 % (> 840 μm) |
| Packaging | Plastic Bottle |
| Water Retention Capacity | 74 to 82 % (H form) |
| Color | Light Beige to Orange to Brown |
| Quantity | 100 g |
| Solubility Information | Insoluble in water |
| Show More |
Safety and Handling
| Hazard Category | Corrosion |
| Recommended Storage | Normal conditions |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction