Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ AmberChrom™ (formerly Dowex™) 50WX2, Hydrogen Form, 100-200 mesh

Catalog No. 111015221
Click to view available options
Quantity:
100 g
2.5 kg

A preferred strong cationic exchange (SCX) resin due to its attractive features such as high capacity, easy availability and ease of operation owing to the lower degree of divinylbenzene cross-linking leading to higher recoveries of aromatic amino acids.

Dowex™ 50WX2 hydrogen form is a heat stable, water stable and low molecular weight cationic substance which is capable of purifying approximately 5000-fold by ion-exchange chromatography. Ion-exchange resins have good role as drug delivery systems.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 100/200 Mesh
Packaging Poly Bottle
pH 0 to 14
Quantity 2.5 kg
Product Line Amberchrom
Capacity (Metric) 0.6 meq/mL by wetted bed volume
MDL Number MFCD00132722
CAS 69011-20-7

Amberchrom is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.