Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite 200, MP Biomedicals™

Catalog No. ICN15027890 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 g
500 g

Strongly acidic macroreticular-type cation exchange resin. Sodium form sulfonic acid functionality

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12626-25-4
MDL Number MFCD00145784

Specifications

Physical Form Powder
Color Yellow
Quantity 500 g
Chemical Name or Material Amberlite
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.