Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amberlite™ HPR650 H, Hydrogen Form

Catalog No. p-7191700
Click to view available options
Quantity:
1 kg

Formerly Dowex™ Monosphere™ 650 C hydrogen form

Dowex Monosphere 650C is a strong basic cation exchange gel resin with sulfonic acid as functional group. Dowex Monosphere 650C is a heterogeneous catalyst.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
pH 0 to 14
Quantity 1 kg
Product Line Amberlite
Capacity (Metric) 2.0 meq/mL by wetted bed volume
MDL Number MFCD00212649
CAS 69011-20-7

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.