Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amberlite™ IRA-410 Chloride Form, Matrix styrene/divinylbenzene, 20-25 mesh

Catalog No. p-7191697 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
500 g

Catalog No. 11-101-4979

Supplier: MilliporeSigma™ Supelco™ 216569500G

Only null left
Add to Cart
Add to Cart

Strongly basic (type II) anion exchange resin for water conditioning, removing iodine ions, neutralization of solutions, capturing xanthan and xanthene dyes, etc.; better regeneration efficiency than type I base resin.

Amberlite™ IRA-410, anion exchange resin, is an effective catalyst especially for those reactions involving low catalyst concentrations and relatively low temperature. Amberlite™ IRA-410 is a gel-type resin having styrene-divinyl-benzene skeletal structure.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 20/25 Mesh
Packaging Poly Bottle
pH 0 to 14
Quantity 500 g
Product Line Amberlite
Capacity (Metric) 1.25 meq/mL by wetted bed volume
MDL Number MFCD00132710
CAS 63181-94-2

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.