Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite IRA-410(Cl), ion exchange resin

Catalog No. aaa1773422
Click to view available options
Quantity:
100 g
500 g
2500 g

Catalog No. AAA1773422

Supplier: Thermo Scientific Chemicals A1773422

Only null left
Add to Cart

CAS: 9002-26-0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-26-0
MDL Number MFCD00132710

Specifications

Odor Amine-like
Merck Index 14,382
Quantity 100 g
Chemical Name or Material Amberlite IRA-410(Cl), ion exchange resin
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.