Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite™ IRN-78, ion exchange resin, nuclear grade

Catalog No. p-4824597
Click to view available options
Quantity:
500 g
2 kg

Strongly basic anion exchange resin | CAS: 11128-95-3 | Styrene-DVB

Amberlite™ IRN-78 is widely employed in removing radioactive anions for example 131 Iodine and 133 Iodine from waste systems. It also removes the traces of chloride contamination from reactor coolant systems and controls the boron level in the primary system. At higher pH conditions, it is used to remove anionic impurities from secondary streams in the presence of ammonia, morpholine or other amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11128-95-3
Molecular Formula Styrene-DVB
MDL Number MFCD00145822

Specifications

Physical Form Beads
Color Amber
Quantity 500 g
Solubility Information Insoluble in water.
Grade Nuclear
Chemical Name or Material Amberlite™ IRN-78
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.