Learn More
MilliporeSigma™ Supelco™ Amberlite™ MB-20, Mixed Bed Ion Exchange Resin, Regenerated
Catalog No. p-7191692 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
250 g
1 kg
Amberlite™ MB-20 may be used for the removal of ions from aqueous solutions.
Amberlite™ MB-20 may be used for the removal of ions from aqueous solutions. Uses include primary water chemistry control in once-through systems, ultrapure water production, condensate polishing, in-process and column demineralization, and removal of ionic detergents (such as SDS) from protein samples.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Poly Bottle
Quantity 250 g
Product Line Amberlite
MDL Number MFCD03791384
Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.