Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite™ XAD-4, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
9003-69-4
C10H10
130.19
MFCD00132704
MYRTYDVEIRVNKP-UHFFFAOYSA-N
divinylbenzene,1,2-divinylbenzene,o-divinylbenzene,benzene, diethenyl,benzene, 1,2-diethenyl,divinylbenzen,divinyl benzen,divinyl-benzene,poly divinylbenzene,divinylbenzene dvb
66666
1,2-bis(ethenyl)benzene
C=CC1=CC=CC=C1C=C
9003-69-4
C10H10
130.19
MFCD00132704
MYRTYDVEIRVNKP-UHFFFAOYSA-N
divinylbenzene,1,2-divinylbenzene,o-divinylbenzene,benzene, diethenyl,benzene, 1,2-diethenyl,divinylbenzen,divinyl benzen,divinyl-benzene,poly divinylbenzene,divinylbenzene dvb
66666
1,2-bis(ethenyl)benzene
C=CC1=CC=CC=C1C=C
Ion exchange resin
Additional Information Ponceau dye test: < 1.0
Moisture Holding Capacity moisture holding capacity: 54.0 to 60.0%
Particle Size 2.0 (uniformity coefficient) Max. 0.490 to 0.690mm (harmonic mean size) 5.0% Max. (<0.300mm) 5.0% Max. (>1.180 mm)
Packaging Plastic bottle
Sodium Chloride 20.0 to 32.0 g/l
Color White
Quantity 100g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Amberlite™ XAD-4