Learn More
Alfa Aesar™ Amberlyst™ 15(H), wet, ion exchange resin
Manufacturer:  Alfa Aesar™ 4407930
Quantity 250g
 View more versions of this product
Catalog No. AA4407930
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
39389-20-3
MFCD00145841
Amberlyst™ 15 (H) ion exchange resin, Wet, Styrene-DVB; Strongly acidic macroreticular resin
Specifications
39389-20-3
MFCD00145841
Hygroscopic
Beads
250g
Amberlyst™ 15 (H) ion exchange resin, Wet, Styrene-DVB; Strongly acidic macroreticular resin
Safety and Handling
SDS & Safety Documents
TSCA : TSCA