Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlyst™ A-21, ion exchange resin

Catalog No. p-7023926
Click to view available options
Quantity:
250 g
1000 g

CAS: 9049-93-8

Amberlyst™ A-21, ion exchange resin is used as a heterogeneous catalyst for conversion of carbonyl compounds to oximes by reacting with hydroxylamine hydrochloride. It is an effective catalyst for the Michael addition of nitroalkanes to methyl acrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlyst|r A-21, ion exchange resin is used as a heterogeneous catalyst for conversion of carbonyl compounds to oximes by reacting with hydroxylamine hydrochloride. It is an effective catalyst for the Michael addition of nitroalkanes to methyl acrylate.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and nitric acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9049-93-8
MDL Number MFCD00145842

Specifications

Odor Odorless
Quantity 250 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlyst™ A-21, ion exchange resin
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.