Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlyst™ A-26(OH), ion exchange resin

Catalog No. p-7023616
Click to view available options
Quantity:
250 g
1000 g

CAS: 39339-85-0

Amberlyst™ A-26(OH) is widely utilized to catalyze reactions such as aldol condensation and carbonylation. It is used to remove the anionic transition metal complexes from aqueous and non-aqueous mediums, acids and mercaptans from hydrocarbons and other non-polar solvents. It plays an essential role for the removal of oleic acid from chlorinated hydrocarbons and acids from phenol-acetone solutions. It is actively involved in the determination of cadmium by flow injection atomic absorption spectrometry and as packaging material for column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlyst|r A-26(OH) is widely utilized to catalyze reactions such as aldol condensation and carbonylation. It is used to remove the anionic transition metal complexes from aqueous and non-aqueous mediums, acids and mercaptans from hydrocarbons and other non-polar solvents. It plays an essential role for the removal of oleic acid from chlorinated hydrocarbons and acids from phenol-acetone solutions. It is actively involved in the determination of cadmium by flow injection atomic absorption spectrometry and as packaging material for column chromatography.

Solubility
Insoluble in water.

Notes
Exposure to air absorbs carbon dioxide. Incompatible with strong acids, bases and oxidants.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39339-85-0
MDL Number MFCD00145579

Specifications

Quantity 250 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlyst™ A-26(OH), ion exchange resin
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.