Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ambertec™ UP550 OH Anion Exchange Resin, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022421 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 kg
5 kg
20 kg

Dowex™ Monosphere™ 550A UPW OH type 1 strong base anion exchange resin is recommended for the roughing, intermediate, and polishing ion exchange applications. Low TOC levels in UPW grade are achieved by deliberate functionalization and rinsing.

  • Light color
  • Superior crush strength
  • High (>95%) conversion of exchange sites to the hydroxide (OH) form
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Formerly Dowex™ Monosphere™ 550 A UPW OH
CAS 69011-18-3, 7732-18-5
Particle Size 590 μm (Standard Deviation: ±50)
Volume Total Capacity 1 eq/L
Water Retention Capacity 55% to 65%
Quantity 5 kg
Grade UPW
Concentration or Composition (by Analyte or Components) Trimethylamine functionalized copolymer of styrene and divinylbenzene in the hydroxide form

AmberTec is a trademark of E. I. du Pont de Nemours and Company

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.