Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guardian Manufacturing 6N3032A, Ambidextrous 30 mil Neoprene Glovebox Gloves with 6 in. Port
SDP

Catalog Number 19321141
Click to view available options
Size: 9.75
This item is not returnable. View return policy
This item is not returnable. View return policy

Guardian™ Ambidextrous 30 mil Neoprene Glovebox Gloves with 6 in. Port for radiation application in defense, industrial and nuclear facilities.

  • Constructed of Neoprene
  • Ambidextrous
  • Soft feel
  • Superior dexterity
Certifications/Compliance ISO 9001, USW, Hubzone certified, BERRY, ASTM D412, AGS-G005-2014
Color Black
Hand Ambidextrous
Latex Free Yes
Material Neoprene
Size 9.75
Thickness (English) 30 mil
For Use With (Application) Chemicals and gases
Quantity 1 pair
Length (English) 32 in.
Diameter (English) Cuff 6 in.
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit