Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynabeads™ Oligo(dT)25

Catalog No. p-4924154
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-002 2 mL
61-005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 61-002 Supplier Invitrogen™ Supplier No. 61002
Only null left
Add to Cart
Edge
Add to Cart

Dynabeads™ Oligo(dT)25 mRNA isolation beads specifically target and capture mRNA molecules from virtually any crude sample and eliminate the need to purify total RNA when the desired information-bearing nucleic acid is mRNA.

Dynabeads™ Oligo(dT)25 mRNA isolation beads specifically target and capture mRNA molecules from virtually any crude sample and eliminate the need to purify total RNA when the desired information-bearing nucleic acid is mRNA. Since mRNA comprises only about 1–5% ot total cellular RNA, the isolation of total RNA is not the most efficient way to isolate mRNA. Other technologies designed to purify total RNA yield ∼80% ribosomal RNA and force mRNA to compete with ribosomal RNA, transfer RNA, micro RNA, small nucleolar RNA, and small cytoplasmic RNA for membrane binding. Advantages of Dynabeads™ Oligo(dT)25 beads:

• Fast and gentle procedure yields pure intact mRNA
• Extremely pure mRNA isolation, best choice upstream of cDNA synthesis
• Exquisitely sensitive mRNA isolation enables cDNA synthesis and cDNA library construction from ultra-small starting samples (enables cDNA library construction from a single cell)

How the beads work
The oligo(dT)25-coated Dynabeads™ specifically target and capture the mRNA transcriptome from an extremely wide variety of crude starting samples. Ribosomal RNA, DNA, proteins, and small RNA molecules (such as transfer RNA, micro RNA, and small nucleolar RNA) do not bind to the beads and are discarded. Only polyadenylated RNA species (mRNA) are captured. Isolated mRNA is pure, eliminating the need for ribosomal RNA subtraction or a post-extraction DNase treatment. This column-free system ensures the highest transcriptome recovery:

• Physical mRNA capture on mobile magnetic beads
• Rapid and gentle magnetic handling procedures
• No mRNA lost during high g-force spins
• No mRNA trapped in column membranes during elution

Applications
mRNA is suitable for all downstream molecular applications, including gene cloning, cDNA synthesis, cDNA library construction, RT-PCR, quantitative RT-PCR, RPA (Ribonuclease Protection Assay), subtractive hybridization, primer extension, SAGE, RACE, and others. The Dynabeads™ Oligo(dT)25 mRNA isolation beads are the ideal mRNA purification method prior to cDNA library construction. Use of these beads ensures the highest recovery and enrichment of the transcriptome. These beads capture more of the transcriptome than is possible with methods that integrate a total RNA isolation step upstream of mRNA isolation.

Verastile elution options
Elution can be performed in any volume down to 5 μL. mRNA elution is optional because enzymatic reactions in downstream procedures are not inhibited by presence of Dynabeads™. Additionally, one can perform cDNA synthesis directly on the beads to create a reusable solid-phase cDNA library.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store 2°C to 8°C
2 mL Beads
High-throughput Compatibility High-throughput Compatible
For Use With (Application) RNA Extraction
Product Line DYNAL, Dynabeads
Product Type mRNA Isolation Bead
Quantity 2 mL
Shipping Condition Room Temperature
Ligand Type Oligo-dT
Form Beads in Suspension
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.