Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fast SYBR™ Green Cells-to-CT™ Kit
GREENER_CHOICE

Ambion™ Fast SYBR™ Green Cells-to-CT™ Kit is a fast, easy, and robust method to go directly from cultured cells to real-time PCR results.

$1722.00 - $6030.00

Specifications

Concentration 20X
Content And Storage Store Stop Solution, DNase I and 20X RT Enzyme Mix at -20°C. Store Lysis Solution, 2X SYBR™ RT Buffer, Fast SYBR™ Green PCR Master Mix at 4°C. Store Nuclease-free Water at room temperature.
Detection Method SYBR
Polymerase AmpliTaq Fast DNA Polymerase
Reaction Speed Fast
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
44-029-56
GREENER_CHOICE
Encompass_Preferred
View Documents
Invitrogen™
4402956
100 reactions
Each for $1,722.00
Only null left
Add to Cart
 
44-029-57
GREENER_CHOICE
Encompass_Preferred
View Documents
Invitrogen™
4402957
400 reactions
Each for $6,030.00
Only null left
Add to Cart
 
Description

Description

Ambion Fast SYBRGreen Cells-to-CT™ Kit is a fast, easy, and robust method to go directly from cultured cells to real-time PCR results. A breakthrough cell lysis and RNA stabilization technology eliminates the laborious and time-consuming RNA purification process completely. The kit includes sufficient reagents for 100 reactions. The Fast SYBRGreen Cells-to-CT Kit contains AmpliTaq™ Fast DNA Polymerase, UP, which allows instant hot start, thus minimizing non-specific product formation with fast real-time PCR.

  • Includes optimized reagents to work efficiently, right out of the box
  • Simple, effective Cells-to-CT technology enables sample preparation at room temperature in only 7 minutes, including DNase treatment
  • Validated accuracy, exceptional reproducibility, and maximal sensitivity from 10 to 100,000 cells per sample; results equivalent to those from purified RNA
  • 2-Step qRT-PCR

Extraordinary Value
The kit includes reagents for sample preparation integrated into a complete, optimized gene expression workflow including reverse transcription reagents and a high-performance PCR master mix with SYBRGreen.

Extraordinary Ease
The protocol begins with a simple 7-minute sample preparation procedure. First, cells are washed in PBS, and then lysed for 5 minutes at room temperature; Dnase treatment can be performed concurrently. The lysis reaction is stopped, and the sample is then ready for reverse transcription or storage at -20°C for up to 5 months. Because samples are processed directly in culture plates (96- or 384-well), sample handling and the potential for sample loss or transfer error are minimized, resulting in higher reproducibility. Following sample preparation, a portion of the cell lysate is added to an RT reaction followed by real-time PCR using the included master mix.

Extraordinary Performance
To demonstrate the performance of the kit, replicate experiments were run using either traditional RNA purification and real-time PCR analysis, or the Fast SYBRGreen Cells-to-CT Kit. The kit shows equivalent performance to results obtained using purified RNA for analysis of 10 to 100,000 cells. In addition, the sensitivity, efficiency, and dynamic range were shown to be superior to competitor lysate kits. Samples prepared with the kit have been tested for compatibility on various real-time PCR platforms including those from Applied Biosystems, Roche, and Bio-Rad.

Accessory Product:
The SYBRGreen Cells-to-CT Control Kit (Cat. No. 4402959) contains a Xeno™ RNA control and primer set and is recommended to monitor the efficiency of cell lysis and amplification inhibition in Cells-to-CT reactions.

Order Info

Shipping Condition: Dry ice

Specifications

Specifications

20X
SYBR
Fast
Fewer resources used and less waste, Less hazardous
ROX (Pre-mixed)
5 min.
M-MLV
RT-PCR
Cells
Store Stop Solution, DNase I and 20X RT Enzyme Mix at -20°C. Store Lysis Solution, 2X SYBR™ RT Buffer, Fast SYBR™ Green PCR Master Mix at 4°C. Store Nuclease-free Water at room temperature.
AmpliTaq Fast DNA Polymerase
7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
SYBR Green
Cells-to-CT™, SYBR™
UP (Ultra Pure)
Dry Ice
Built-In Hot Start
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.