Learn More
Invitrogen™ IPTG (Dioxane-Free)

Description
Invitrogen IPTG (Isopropyl-beta-D-thiogalactoside) is an inducer of the lac operon and is frequently used in conjunction with X-GAL to determine the lac phenotype of recombinant plasmids or phage. IPTG is also used to induce the expression of cloned genes that are under control of the lac operon.
Order Info
Shipping Condition: Room temperature

Specifications
Specifications
Chemical Name or Material | IPTG (Isopropyl β-D-thiogalactopyranoside) |
Content And Storage | Store at –20°C. |
Packaging Type | Bottle |
Product Line | Ambion™ |
Quantity | 5 g |
Shipping Condition | Room Temperature |
Form | Powder |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.