Learn More
Invitrogen™ MAXIscript™ SP6/T7 Transcription Kit
Description
Includes
SP6 Enzyme Mix, T7 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, pTRI-b-actin, Nuclease-free Water
Ambion™ MAXIscript™ SP6/T7 In Vitro Transcription Kit synthesizes RNA probes in just 10 minutes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations. Each kit contains sufficient reagents for 15 reactions of each enzyme.
- High yields of high specific activity, full-length RNA probes in just 10 minutes
- Ideal probe synthesis kit for nuclease protection assays, Northern and Southern blots, and in situ hybridization
- Includes TURBO DNase™
- Contain quality-tested DNase/RNase-free reagents
- Synthesizes RNA probes with specific activities reaching 1 x 109 cpm/μg in just 10 minutes
- Effcient at synthesizing full-length probes, even at limiting nucleotide concentrations, making them the perfect companion to ribonuclease protection assays
- For synthesis of nonisotopically labeled probes
- Labeling can be accomplished post-transcriptionally by using Ambion's BrightStar™ Psoralen-Biotin Labeling Kit, or during transcription by incorporating modified nucleotides into reaction
- Used for synthesis of modest amounts of unlabeled RNA (2–6μg/20μL reaction) of up to 2kb
Specifications
Specifications
| Content And Storage | SP6 Enzyme Mix, T7 Enzyme Mix,10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, and pTRI-b-actin are all stored at –20°C. Nuclease-free Water may be stored at any temperature. |
| Final Product Type | Synthetic mRNA, mRNA |
| Format | Kit |
| Sample Type | mRNA, Synthetic mRNA |
| For Use With (Application) | In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products |
| No. of Reactions | 100 Reactions |
| Product Line | Ambion, MAXIscript |
| Product Type | SP6/T7 In Vitro Transcription kit |
| Quantity | 100 Reactions |
| Promoter | T7, SP6 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.