Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mRNA Catcher™ PLUS Purification Kit

Catalog No. p-4918729
Encompass_Preferred
Click to view available options
Quantity:
96 Preps
960 Preps
Product Type:
Buffers Only
Purification Kit

Includes

1 96-well mRNA Catcher PLUS plate; 4mL 2X Lysis Buffer L20; 30mL Wash Buffer W15; 4mL Dilution Buffer DL1; 8mL Elution Buffer E3; 100μL 0.5M DTT (4°C)

The mRNA Catcher PLUS Purification Kit enables high-throughput purification of mRNA from cells, tissues, blood, and total RNA in a 96-well format.

The mRNA Catcher PLUS Purification Kit enables high-throughput purification of mRNA from cells, tissues, blood, and total RNA in a 96-well format. By taking advantage of locked nucleic acid (LNA) technology, the mRNA Catcher PLUS kit provides improved performance and a faster protocol, while preserving mRNA yield and purity.

The mRNA Catcher PLUS Purification Kit offers:

  • Robust performance—high binding capacity per well (>80 ng/well)
  • Reproducible results—low CVs from well-to-well and plate-to-plate (∼2%)
  • Optimized protocols—for small sample sizes (100 cells, 4 mg tissue, and 40 μL blood)
  • Compatible plate design—may be used with most PCR cyclers

How it works

mRNA from total RNA, cells, tissue, and blood samples is hybridized to an immobilized, single-stranded 20-mer oligo(dT)-containing LNA. The incorporation of LNA into the oligo probe increases the hybridization efficiency and specificity of the mRNA binding. The mRNA can either be eluted or reversed-transcribed to cDNA directly in the well. The mRNA Catcher PLUS method does not require centrifugation, precipitation, phase separation, or column filtration. In addition, the entire method is amenable to automation for streamLined, high-throughput expression profiling for large-scale processes. Isolated mRNA is suitable for use in downstream applications such as RT-PCR and qRT-PCR.

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 x 96-well mRNA Catcher™ PLUS Plate; store at 4°C
• 4 mL 2X Lysis Buffer L20; room temperature
• 30 mL Wash Buffer W15; room temperature
• 4 mL Dilution Buffer DL1; room temperature
• 8 mL Elution Buffer E3 (DEPC-treated RNase-free water); room temperature
• 100 μL 0.5M DTT; 4°C
Isolation Technology Oligo dT Affinity
Purification Time 1.5 hr. (Including sample homogenization)
Elution Volume 80 μL
Sample Type Cells, Tissue, Total RNA, Whole Blood
Final Product Type mRNA
High-throughput Compatibility High-throughput Compatible
For Use With (Equipment) Robotic Liquid Handling Systems
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, Microarray Analysis
Product Type Purification Kit
Quantity 96 Preps
Shipping Condition Room Temperature
Starting Material Amount Cells: 100–106, Tissue: 4–40 mg, Total RNA: 0.1–100 μg, Whole Blood: 40 μL
Yield >80 ng
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.