Learn More
Invitrogen™ RNAqueous™ Total RNA Isolation Kit

Description
The RNAqueous Total RNA Isolation Kit is for phenol-free total RNA isolation from 1–75 mg of tissue or from 102–107 cells, using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.
The RNAqueous kit provides:
- Convenient syringe-based filtration
- Compatibility with vacuum manifolds when processing multiple samples
How it works
The RNAqueous kit uses a powerful chaotropic solution for cell lysis, followed by RNA binding to glass fiber housed in a luer-lock syringe filter. Washing and elution solutions are simply pushed through the filter using a standard syringe (not included). Vacuum filtration is not required; however, the filter units are compatible with standard vacuum manifolds for processing several samples at once.
Order Info
Shipping Condition: Room temperature

Specifications
Specifications
Content And Storage | 50 cartridges; store at room temperature. |
For Use With (Application) | RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling |
Product Type | Spin Columns |
Quantity | 50 Catridges |
Shipping Condition | Room Temperature |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.