Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tris (1 M), pH 8.0, RNase-free

Catalog No. p-4926942
Encompass_Preferred
Click to view available options
Quantity:
100 mL
500 mL

Invitrogen Molecular biology grade, 1M Tris, pH 8.0 solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use.

Invitrogen Molecular biology grade, 1M Tris, pH 8.0 solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 M
pH 8
Physical Form Liquid
Product Line Ambion
Quantity 100 mL
Formulation 1,3-Propanediol, 2-Amino-2-(Hydroxymethyl)-,Hydrochloride
Grade Molecular Biology
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.