Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ULTRAhyb™ Ultrasensitive Hybridization Buffer

Catalog No. p-4926722
Encompass_Preferred
Change view
Click to view available options
Quantity:
125 mL
4 x 125 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8670 125 mL
AM8669 4 x 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM8670 Supplier Invitrogen™ Supplier No. AM8670
Only null left
Add to Cart
Edge
Add to Cart

Proprietary blend of hybridization accelerators and blocking agents that greatly enhance the levels of hybridization

  • Signals that once took days to visualize are now apparent in hours
  • It is supplied in four bottles containing 125mL each
  • Increase the sensitivity of blot hybridization experiments
  • Increase random-primed DNA probe sensitivity up to 100x; RNA probe sensitivity 20x
  • Decrease hybridization time to 2 hours
  • Using standard hybridization buffers, only 1 to 5% of target molecules on a blot hybridizes to probe, making blots a relatively insensitive method for nucleic acid analysis (Vernier et al. (1996) Anal Biochem 235: 11-19); With ULTRAhyb Ultrasensitive Hybridization Buffer, the hybridization reaction approaches completion, so that as few as 10,000 molecules can be detected
  • Compatible with RNA and DNA probes labeled isotopically and nonisotopically in northerns, Southerns, and dot/slot blots
  • Contains 50% formamide and is compatible with positively charged membranes
  • Requires 10-100x less nonisotopic DNA probe than standard hybridization buffers, making it possible to perform many more hybridizations with a single labeling reaction

Applications:

DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C.
Buffer Hybridization Buffer
Quantity 125 mL
Shipping Condition Room Temperature
Product Type Ultrasensitive Hybridization Buffer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.