Ambion™ UltraPure™ Buffer-Saturated Phenol Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Buffer for nucleic acid purification

Manufacturer: Invitrogen 15513047

 View more versions of this product

Catalog No. 15-513-047

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Quantity 400mL
    Storage Requirements Store in refrigerator (2° to 8°C).

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit