Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMBRA1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
| Antigen | AMBRA1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224723SS
![]() |
Novus Biologicals
NBP224723SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP224723
![]() |
Novus Biologicals
NBP224723 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
AMBRA1 Polyclonal specifically detects AMBRA1 in Human, Mouse, Rat, Bovine, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| AMBRA1 | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Bovine, Chicken | |
| Q9C0C7 | |
| 55626 | |
| A portion of amino acids 1-50 of human Ambra1 was used as the immunogen for the antibody. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Autophagy | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| activating molecule in BECN1-regulated autophagy protein 1, autophagy/beclin-1 regulator 1, DCAF3, DDB1 and CUL4 associated factor 3, FLJ20294, KIAA1736activating molecule in beclin-1-regulated autophagy, MGC33725, WD repeat domain 94, WDR94 | |
| AMBRA1 | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen for the antibody is 100% homologous in human, zebra fish, wild boar, duck billed platypus, rat, mouse, chimpanzee, opossum, macaques, monkey, dog, xenopus and chicken. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title