Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMBRA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520512
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520512 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520512 Supplier Invitrogen™ Supplier No. PA520512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-20512 can be used with blocking peptide PEP-0632.

Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. Beclin-1, a principal regulator of autophagosome formation, is in turn regulated by Ambra1. Ambra1 associates with Beclin-1 through a region near its center as determined by yeast two-hybrid assay. Null mutations in this gene in mice resulted in embryonic lethality with severe neural tube defects associated with autophagy impairment, accumulation of ubiquitinated proteins, unbalanced cell proliferation and excessive apoptotic death. Furthermore, down-regulation of Ambra1 in cultured cells though RNA interference decreased the level of rapamycin- and nutrient starvation-induced autophagy. Multiple isoforms of Ambra1 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMBRA1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene AMBRA1
Gene Accession No. A2AH22, Q9C0C7
Gene Alias 2310079H06Rik; A130023A14; AA474864; activating molecule in beclin-1-regulated autophagy; activating molecule in Beclin1-regulated autophagy; activating molecule in BECN1-regulated autophagy protein 1; AMBRA1; autophagy and beclin 1 regulator 1; autophagy/beclin 1 regulator 1; autophagy/beclin-1 regulator 1; AV021921; D030051N19Rik; DCAF3; DDB1 and CUL4 associated factor 3; hypothetical protein; hypothetical protein LOC517263; ischemia related factor NYW-1; Kiaa1736; mKIAA1736; Nyw1; WD repeat domain 94; WDR94
Gene Symbols AMBRA1
Host Species Rabbit
Immunogen An 18 amino acid peptide from near the amino terminus of human Ambra1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228361, 55626, 59319
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.