Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMBRA1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501562908
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2908 Supplier Bethyl Laboratories, Inc Supplier No. A302568A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Ambra1, a WD40 domain containing protein, is a novel activating molecule in Beclin 1-regulated autophagy and plays a role in the development of the nervous system. The protein binds to Beclin 1 and favors Beclin 1/Vps34 interaction. The Ambra1 gene localized on human chromosome 11p11.2, encodes the protein that is highly conserved in vertebrates only and its expression is mostly confined to the neuroepithelium during early neurogenesis.

Specifications

Antigen AMBRA1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene AMBRA1
Gene Accession No. Q9C0C7
Gene Alias DCAF3, WDR94; DDB1 and CUL4 associated factor 3; WD repeat domain 94; activating molecule in BECN1-regulated autophagy protein 1; activating molecule in beclin-1-regulated autophagy
Gene Symbols AMBRA1
Host Species Rabbit
Immunogen Between 1200 and 1250
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55626
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.