Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMBRA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501728990 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8990 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8990 Supplier Proteintech Group Inc Supplier No. 137621AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody works well in WB, IHC and IF application.

AMBRA1 (activating molecule in Beclin-1-regulated autophagy protein 1) is a protein that bears a WD40 domain and regulates Beclin-1-dependent autophagy. In mouse embryos AMBRA1 has been shown to be critical for neural tube development.

Specifications

Antigen AMBRA1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AMBRA1
Gene Accession No. A2AH22, Q9C0C7
Gene Alias AMBRA1, autophagy/beclin 1 regulator 1, KIAA1736, WDR94
Gene Symbols Ambra1
Host Species Rabbit
Immunogen AMBRA1 Fusion Protein Ag4735
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228361, 55626, 59319
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.