Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ambroxol Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A236025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A236025G 25 g
A23605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A236025G Supplier TCI America Supplier No. A236025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23828-92-4
Molecular Formula C13H19Br2ClN2O
Molecular Weight (g/mol) 414.57
MDL Number MFCD00078932
InChI Key QNVKOSLOVOTXKF-UHFFFAOYSA-N
Synonym ambroxol hydrochloride, ambroxol hcl, mucoangin, mucosolvan, ambroxolhydrochloride, unii-cc995zmv90, cis-4-2-amino-3,5-dibromobenzyl amino cyclohexanol hydrochloride, trans-4-2-amino-3,5-dibromobenzyl amino cyclohexanol hydrochloride, ambroxol hydrochloride jan, lasolvan
PubChem CID 108013
IUPAC Name hydrogen 4-{[(2-amino-3,5-dibromophenyl)methyl]amino}cyclohexan-1-ol chloride
SMILES [H+].[Cl-].NC1=C(Br)C=C(Br)C=C1CNC1CCC(O)CC1

Specifications

Melting Point 235°C
Color White
pH 4.0 to 6.0 (1% H2O)
Quantity 25 g
Formula Weight 414.57
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ambroxol Hydrochloride
RTECSNumber GV8425000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.