Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AmCyan-Tag Monoclonal Antibody (5F9), Biotin, Bioss

Mouse Monoclonal Antibody
Supplier: Bioss BSM33184MBIOTIN

Description
AmCyan-Tag Monoclonal antibody specifically detects AmCyan-Tag in Tag samples. It is validated for Western BlotSpecifications
AmCyan-Tag | |
Monoclonal | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
Mouse | |
Protein G | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Western Blot | |
5F9 | |
Biotin | |
AmCyan1; cyan fluorescent | |
Recombinant AmCyan | |
100 μL | |
Primary | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction