Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544172
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544172 Supplier Invitrogen™ Supplier No. PA544172
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MGRMNSDCWY LYTLDFPESR VISQPDQTLE ILMSELDPAV MDQFYMKDGV Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

This gene encodes an important intermediate enzyme in polyamine biosynthesis. The polyamines spermine, spermidine, and putrescine are low-molecular-weight aliphatic amines essential for cellular proliferation and tumor promotion. Two alternatively spliced transcript variants that encode different proteins have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AMD1
Gene Accession No. P17707
Gene Alias 1; adenosylmethionine decarboxylase 1; AdoMet decarboxylase; adoMetDC; AdoMetDC 1; adoMetDC1; AMD; Amd1; Amd-1; Amd1a; Amd1b; DKFZp313L1234; FLJ26964; S-adenosylmethionine decarboxylase; S-adenosylmethionine decarboxylase 1; S-adenosylmethionine decarboxylase 1 alpha chain; S-adenosylmethionine decarboxylase 1 beta chain; S-adenosylmethionine decarboxylase 1A; S-Adenosylmethionine decarboxylase 1B; S-adenosylmethionine decarboxylase alpha chain; S-adenosylmethionine decarboxylase beta chain; S-adenosylmethionine decarboxylase proenzyme; S-adenosylmethionine decarboxylase proenzyme 1; S-adenosylmethionine decarboxylase proenzym-like protein; SAMDC; SAMDC 1
Gene Symbols AMD1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human AMD1 (aa 149-198).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 262
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.