Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMD1 Polyclonal Antibody
AMD1 Polyclonal Antibody
Supplier: Thermo Scientific PA583077
Description
AMD1 Polyclonal Antibody for ICC/IF, IHC (P)

Specifications
AMD1 | |
Polyclonal | |
Unconjugated | |
AMD1 | |
1; adenosylmethionine decarboxylase 1; AdoMet decarboxylase; adoMetDC; AdoMetDC 1; adoMetDC1; AMD; Amd1; Amd-1; Amd1a; Amd1b; DKFZp313L1234; FLJ26964; S-adenosylmethionine decarboxylase; S-adenosylmethionine decarboxylase 1; S-adenosylmethionine decarboxylase 1 alpha chain; S-adenosylmethionine decarboxylase 1 beta chain; S-adenosylmethionine decarboxylase 1A; S-Adenosylmethionine decarboxylase 1B; S-adenosylmethionine decarboxylase alpha chain; S-adenosylmethionine decarboxylase beta chain; S-adenosylmethionine decarboxylase proenzyme; S-adenosylmethionine decarboxylase proenzyme 1; S-adenosylmethionine decarboxylase proenzym-like protein; SAMDC; SAMDC 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
262 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P17707 | |
AMD1 | |
Recombinant protein corresponding to Human AMD1. Recombinant protein control fragment (Product #RP-94737). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction