Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AMDHD1 Monoclonal Antibody (OTI4C1)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527240
Description
AMDHD1 Monoclonal Antibody for Western Blot, IHC (P)
AMDHD1 is a protein coding gene.
Specifications
AMDHD1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96NU7 | |
AMDHD1 | |
Full length human recombinant protein of AMDHD1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4C1 | |
Unconjugated | |
AMDHD1 | |
1300019J08Rik; Amdhd1; amidohydrolase domain containing 1; amidohydrolase domain-containing protein 1; HMFT1272; Probable imidazolonepropionase; RGD1308879 | |
Mouse | |
Affinity chromatography | |
RUO | |
144193 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction