Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMDHD2 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299173MFV500
Description
AMDHD2 Polyclonal antibody specifically detects AMDHD2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
AMDHD2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
amidohydrolase domain containing 2, Amidohydrolase domain-containing protein 2, CGI-14, EC 3.5.1.25, GlcNAc 6-P deacetylase, putative N-acetylglucosamine-6-phosphate deacetylase | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human AMDHD2. | |
0.1 mL | |
Lipid and Metabolism | |
51005 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction