Learn More
Invitrogen™ AMDHD2 Polyclonal Antibody

Description
Immunogen sequence: FITHLFNAML PFHHRDPGIV GLLTSDRLPA GRCIFYGMIA DGTHTNPAAL RIAHRAHPQG LVLVTDAIPA LGLGNGRHTL GQQEVEVDGL T Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Specifications
Specifications
| Antigen | AMDHD2 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | AMDHD2 |
| Gene Accession No. | Q9Y303 |
| Gene Alias | 5730457F11Rik; AMDHD2; amidohydrolase domain containing 2; amidohydrolase domain-containing protein 2; CGI-14; GlcNAc 6-P deacetylase; N-acetylglucosamine-6-phosphate deacetylase; putative N-acetylglucosamine-6-phosphate deacetylase |
| Gene Symbols | AMDHD2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.