Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ameloblastin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567953
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567953 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567953 Supplier Invitrogen™ Supplier No. PA567953
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Human samples.

This gene encodes the nonamelogenin enamel matrix protein ameloblastin. The encoded protein may be important in enamel matrix formation and mineralization. This gene is located in the calcium-binding phosphoprotein gene cluster on chromosome 4. Mutations in this gene may be associated with dentinogenesis imperfect and autosomal dominant amylogenesis imperfect.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ameloblastin
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AMBN
Gene Accession No. Q9NP70
Gene Alias AI1F; AMBN; Amelin; ameloblastin; ameloblastin (enamel matrix protein); ameloblastin precursor; enamel matrix protein
Gene Symbols AMBN
Host Species Rabbit
Immunogen Synthesized peptide derived from human Ameloblastin.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 258
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.